Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Doxorubicin Hydrochloride Inhibitor
SDP

Catalog No. NB225250MG Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB225250MG 50 mg
NB225210MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB225250MG Supplier Novus Biologicals™ Supplier No. NB225250MG

missing translation for 'validMainframeMsg'

Low molecular weight, organic compound that may help regulate biological process.

Biological Activity: Antitumor antibiotic agent that inhibits DNA topoisomerase II. DNA intercalator that inhibits nucleic acid synthesis and induces apoptosis. Reduces intracellular tau levels.

Storage: Store at 4°C.

Note: Inhibitors are guaranteed for 1 year from date of receipt.

Specifications

Color Orange
Product Type Inhibitor
Target Proteins Doxorubicin hydrochloride
CAS 25316-40-9
Molecular Formula C30H41FN4O12
Formula Weight 579.99g/mol
Solubility Soluble to 50nM in DMSO, 50nM water
Content And Storage Store at 4°C
For Use With (Application) Inhibits apoptosis
Quantity 50 mg

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.