Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Doxorubicin Hydrochloride, Topoisomerase II Inhibitor, Solid, 10 mg

Catalog No. 502584584
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-4584 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-4584 Supplier Abcam Supplier No. AB12062910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent DNA intercalating agent and Topoisomerase II inhibitor that induces apoptosis.

  • Purity: >98%
  • Molecular Weight: 580 Da
  • Mechanism of Action: Topoisomerase II inhibitor (IC50 = 100 nM, Hep3B cells); DNA intercalator that prevents synthesis of nucleic acid
  • Biological Effect: Induces apoptosis
  • CAS Number: 25316-40-9
  • Form: Solid

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Topoisomerase II, DNA
Content And Storage Store at -20°C.
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Form Solid
Formulation 100% Doxorubicin hydrochloride
Quantity 10 mg
For Use With (Application) Topoisomerase II inhibition, DNA intercalation, Apoptosis induction
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.