Learn More
Abcam Doxorubicin Hydrochloride, Topoisomerase II Inhibitor, Solid, 50 mg
Description
- Molecular Weight: 580 Da
- Purity: >98%
- Form: Solid
- Mechanism of Action: Topoisomerase II inhibitor (IC50 = 100 nM, Hep3B cells); DNA intercalator that prevents synthesis of nucleic acid; Induces apoptosis
- Storage Conditions: Store at -20°C
- Shipping Conditions: Ambient - Can Ship with Ice
- Citations: 13
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Target Molecule | DNA, Topoisomerase II |
| Formulation | 100% Doxorubicin hydrochloride |
| Quantity | 50 mg |
| For Use With (Application) | Topoisomerase II inhibition, DNA intercalation, apoptosis induction |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.