Learn More
Abcam Doxycycline Hyclate, Matrix Metalloprotease Inhibitor, >97% Purity, 1 g
Description
- Purity: >97%
- Molecular Weight: 1,025.9 Da
- Form: Solid
- Function: Synthetic tetracycline; matrix metalloprotease (MMP) inhibitor
- Biological activity: Inhibits tumour growth, suppresses cell proliferation and induces apoptosis in vitro and in vivo
- Storage: Store at +4°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Buffer | 100% Doxycycline hyclate |
| Content And Storage | Store at 4°C. |
| Form | Solid |
| Target Molecule | Matrix Metalloprotease |
| Formulation | 100% Doxycycline hyclate |
| Quantity | 1 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.