Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Doxycycline Hyclate, Matrix Metalloprotease Inhibitor, >97% Purity, 5 g
Description
- Chemical class: Synthetic tetracycline
- Biological activity: Matrix metalloprotease (MMP) inhibitor; inhibits tumour growth, suppresses cell proliferation, and induces apoptosis in vitro and in vivo
- Purity: >97%
- Molecular weight: 1025.9 Da
- Form: Solid
- CAS number: 24390-14-5
- Storage: Store at +4°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Matrix Metalloproteases |
| Buffer | 100% Doxycycline hyclate |
| Content And Storage | Store at 4°C. |
| Form | Solid |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Formulation | 100% Doxycycline hyclate |
| Quantity | 5 g |
| For Use With (Application) | Tumour growth inhibition, cell proliferation suppression, apoptosis induction |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?