Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Doxycycline Hyclate, Matrix Metalloprotease Inhibitor, Solid, 10 g

Catalog No. 502583370
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3370 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3370 Supplier Abcam Supplier No. AB14109110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic tetracycline and matrix metalloprotease inhibitor that suppresses cell proliferation and induces apoptosis.

  • Purity: >97%
  • Molecular Weight: 1025.9 Da
  • Mechanism: Synthetic tetracycline and matrix metalloprotease (MMP) inhibitor
  • Biological Activity: Inhibits tumour growth, suppresses cell proliferation and induces apoptosis in vitro and in vivo
  • Form: Solid
  • Storage: Store at +4°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Form Solid
Content And Storage Store at 4°C.
Target Molecule Matrix Metalloprotease
Shipping Condition Ambient
Formulation 100% Doxycycline hyclate
Quantity 10 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.