Learn More
Abcam Doxycycline Hyclate, Matrix Metalloprotease Inhibitor, Solid, 25 g
Description
- Chemical Type: Synthetic tetracycline
- Biological Activity: Matrix metalloprotease (MMP) inhibitor; inhibits tumour growth, suppresses cell proliferation, and induces apoptosis in vitro and in vivo
- Purity: >97%
- Molecular Weight: 1,025.9 Da
- Storage Temperature: Store at +4°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Matrix Metalloproteinase |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Content And Storage | Store at 4°C. |
| Form | Solid |
| Formulation | 100% Doxycycline hyclate |
| Quantity | 25 g |
| For Use With (Application) | Matrix metalloprotease inhibition, tumour growth inhibition, cell proliferation suppression, apoptosis induction |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.