Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Doxycycline hyclate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012182 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2182 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2182 Supplier MilliporeSigma Supelco Supplier No. PHR11451G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Doxycycline, the semi-synthetic analog of tetracycline, is a broad-spectrum antibiotic employed in medicine and veterinary applications, is known for its action against a huge range of gram positive and gram negative bacteria. It is prescribed against infections spread by rickettsiae, mycoplasmas and chlamydiae. Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 24390-14-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C22H24N2O8 · HCl · 0.5H2 O · 0.5C2H6 O
MDL Number MFCD07357237
Synonym Doxycycline hydrochloride hemiethanolate hemihydrate
Type Certified Reference Material
Quantity 1 g
Formula Weight 512.94
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.