Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Doxycycline hydrochloride, 20 mg/ml in distilled water, sterile-filtered

Catalog No. AAJ67043AD
Change view
Click to view available options
Quantity:
50 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67043AD 50 mL
AAJ67043AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67043AD Supplier Thermo Scientific Chemicals Supplier No. J67043AD
Add to Cart

Doxycycline hydrochloride, CAS # 10592-13-9, is the hydrochloride salt form of doxycycline that shows antibacterial and antimalarial activities. | CAS: 10592-13-9 | C22H25ClN2O8 | 480.90 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

General Description

• Doxycycline hydrochloride is a compound that shows antibacterial and antimicrobial activities against several species, including Pseudomonas aeruginosa, Plasmodium falciparum, and Escherichia coli. It is also a bacteriostatic

• It can block the binding of aminoacyl-tRNA to the mRNA-ribosome complex, thereby inhibiting protein synthesis

Application

• It is used to eliminate Borrelia burgdorferi and Anaplasma phagocytophilum in rodent reservoirs and eliminate Ixodes scapularis ticks

• Doxycycline hydrochloride is also considered to be a broad-spectrum inhibitor used to inhibit matrix metalloproteinases (MMP), such as type 1 collagenase, in wound healing and tissue remodeling studies. Due to this activity, it can inhibit angiogenesis and reduce lung metastases

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10592-13-9
Molecular Formula C22H25ClN2O8
Molecular Weight (g/mol) 480.90
MDL Number MFCD03427564
InChI Key VLUQVUWDECWBTL-UQVCFKGQSA-N
PubChem CID 54706018
IUPAC Name (1S,3Z,4aS,11R,11aR,12S,12aR)-3-[amino(hydroxy)methylidene]-4a,6,7,12-tetrahydroxy-N,N,11-trimethyl-2,4,5-trioxo-1,2,3,4,4a,5,11,11a,12,12a-decahydrotetracen-1-aminium chloride
SMILES [Cl-].C[C@@H]1[C@H]2[C@H](O)[C@H]3[C@H]([NH+](C)C)C(=O)\C(=C(/N)O)C(=O)[C@@]3(O)C(=O)C2=C(O)C2=C(O)C=CC=C12

Specifications

Type Doxycycline Hydrochloride
Physical Form Liquid
Concentration 20mg/mL in distilled water
Quantity 50 mL
Solubility Information Soluble in water.
Chemical Name or Material Doxycycline hydrochloride, sterile-filtered
Hazard Category H335
Hazard Statement GHS H Statement
H350
May cause cancer.
Precautionary Statement GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 234-198-7
TSCA No
storageNote1 −30° to −10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.