Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Doxylamine Succinate, USP, 98-101%, Spectrum™ Chemical
SDP

Catalog No. 18602335 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-335 5 g Amber Glass Bottle
18-602-336 25 g Amber Glass Bottle
18-602-337 100 g Amber Glass Bottle
18-602-338 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-602-335 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. DO1245GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

DO124, 562-10-7, C17H22N2O·C4H6O4

Spectrum™ Chemical Doxylamine Succinate, USP is an antihistamine, used to relieve symptoms of allergy, hay fever, and the common cold. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 562-10-7
Molecular Formula C21H28N2O5
Molecular Weight (g/mol) 388.46
MDL Number MFCD00056168
InChI Key KBAUFVUYFNWQFM-UHFFFAOYNA-N
IUPAC Name butanedioic acid; dimethyl({2-[1-phenyl-1-(pyridin-2-yl)ethoxy]ethyl})amine
SMILES OC(=O)CCC(O)=O.CN(C)CCOC(C)(C1=CC=CC=C1)C1=CC=CC=N1

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.5%
Melting Range Within 3°C
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 98 to 101%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.