Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103128
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103128 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103128 Supplier Invitrogen™ Supplier No. PA5103128
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DP-2.

The gene is a member of the transcription factor DP family. The encoded protein forms heterodimers with the E2F transcription factors resulting in transcriptional activation of cell cycle regulated genes. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DP2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TFDP2
Gene Accession No. Q14188, Q64163
Gene Alias 1110029I05Rik; A330080J22Rik; BTPS2; DP2; DP2.5; DP3; DP-3; E2F dimerization partner 2; GS; Tcfdp2; TFDP2; transcription factor Dp 2; transcription factor Dp-2; transcription factor Dp-2 (E2F dimerization partner 2); Unknown (protein for MGC:133813)
Gene Symbols TFDP2
Host Species Rabbit
Immunogen A synthesized peptide derived from human TFDP2(Accession Q14188), corresponding to amino acid residues A95-R145.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 211586, 300947, 7029
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.