Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech Dulbecco's Phosphate-Buffered Saline (DPBS), w/o: Ca and Mg (1000 ml)
SDP
Supplier:  ILEX LIFE SCIENCES LLC P04361000
Encompass_Preferred
DPBS, WITHOUT: Ca and Mg. QUANTITY: 1000 ml. MANUFACTURED BY: Pan-Biotech (Germany). STORAGE: 2 to 8 degrees Celsius. DESCRIPTION: Dulbecco's Phosphate-Buffered Saline (DPBS) without calcium chloride and magnesium chloride. The main purpose of buffered salt solutions is the maintenance of pH and osmolarity in cell culture applications as well as providing essential inorganic ions. These solutions are usually used for washing tissues and cells. Often they are combined with other agents in cell and tissue treatment.
Catalog No. 50-238-8549
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.