Learn More
TARGETMOL CHEMICALS INC DPCPX 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T22744
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DPCPX (PD 116948) is an A1 adenosine receptor antagonistpurity: 99%
Catalog No. 50-258-0928
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.