Learn More
LaMotte™ DPD Chlorine Secondary Standards Kit for LaMotte Colorimeters
Shop All LaMotte Company Products
Description
- Use once meter has been calibrated with a primary standard
- Can be inserted periodically for a quick check of instrument calibration
Specifications
Specifications
| For Use With (Application) | Use once meter has been calibrated with a primary standard |
| For Use With (Equipment) | LaMotte colorimeters |
| Includes | Blank and three standards, each with certificate of analysis, for low, mid, and high-range chlorine calibrations, packaged in a plastic case |
| Quantity | Each |
| Type | DPD Chlorine Secondary Standards Kit |