Learn More
Hach Company DPD Free Chlorine Reagent Powder Pillows, 10 mL, pk/1000
Supplier: Hach Company 2105528
For free chlorine determination by the DPD Free Chlorine method. USEPA accepted for reporting for drinking water analysis using Hach Method 8021. Range: 0.02 - 2.00 mg/L Cl₂. Sample Size: 10 mL. Pack of 1000 powder pillows.Boric Acid free!*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.