Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPD Indicator Solution For Residual Chlorine Analysis, Reagents

Catalog No. p-200022042
Click to view available options
Quantity:
100 mL
1 L
4 L
Packaging:
Glass Bottle
Opaque HDPE

DPD Indicator Solution For Residual Chlorine Analysis, Reagent, for general laboratory use. Recommended Storage: 2°C to 8°C (36°F to 46°F).Manufactured in ISO 9001 facility.

Specifications

Color Colorless to Light Pink
Volume (Metric) 100 mL
Packaging Glass Bottle
Quantity 100 mL
Sterility Non-sterile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.