Learn More
Description
With over 50 years of manufacturing water quality reagents, Lovibond™ knows choosing the right test for your process or application is critical for accurate results. Each tablet is protected from humidity, heat and other environmental interferences, as pharmaceutical-grade aluminum blister packaging individually seals up to 10 tablets on an individual strip.
- Easy to dispense blister pack makes dispensing tablets safer
- Shelf-stable, analytical solution for testing water quality parameters
- Accurate and precise dosing
- Long shelf-life
Specifications
Specifications
| Analytes | Chlorine |
| Format | Tablet |
| For Use With (Application) | Water Quality Analysis |
| For Use With (Equipment) | Lovibond™ Colorimeters and Spectrophotometers |
| No. of Tests | 250 Tests |
| Sample Size | 10 mL |
| Test Method | DPD |
| Test Time | 2 min |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.