Learn More
Cayman Chemical ANTIBODY t3 25mg
SDP
Supplier:  Cayman Chemical 3790925 MG
Encompass_Preferred
An inhibitor of microtubule assembly (EC50 3 UM) reduces viability of HeLa cells (EC50 0.53 UM) induces genotoxic stress in a reporter assay using HEK293 cells reduces but does not eradicate worm burden in goats with naturally acquired or experimental nematode infections at 15 mg/kg teratogenic to lamb embryos when administered to pregnant ewes at 60 mg/kg during the third week of pregnancy
Catalog No. 50-291-5555
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.