Learn More
CPC Scientific H-Tyr-DPen-Gly-Phe-Pen-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific ENKP002A
Encompass
SEQUENCE: H-Tyr-DPen-Gly-Phe-Pen-OH (trifluoroacetate salt)(Pen2 and Pen5 bridge)ONE-LETTER SEQUENCE: Y-DPen-GF-Pen(Pen&Pen bridge)MOLECULAR FORMULA: C30H39N5O7S2MOLECULAR WEIGHT: 645.8STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [88373-72-2]SYNONYMS: RESEARCH AREA: PainREFERENCES:H.I. Mosberg et al., PNAS, 80, 5871 (1983)SKU(s): ENKP-002A, ENKP-002B
Catalog No. 50-210-9250
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.