Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DPEP1 Monoclonal Antibody (C1), Invitrogen™

Catalog No. p-7240970
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543850 20 μL
PIMA543849 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543850 Supplier Invitrogen™ Supplier No. MA543850
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

The protein encoded by this gene is a kidney membrane enzyme involved in the metabolism of glutathione and other similar proteins by dipeptide hydrolysis. The encoded protein is known to regulate leukotriene activity by catalyzing the conversion of leukotriene D4 to leukotriene E4. This protein uses zinc as a cofactor and acts as a disulfide-linked homodimer. Two transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPEP1
Applications Western Blot
Classification Monoclonal
Clone C1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene DPEP1
Gene Accession No. P16444
Gene Alias AI327012; Dehydropeptidase-I; dipeptidase 1; dipeptidase 1 (renal); Dpep1; hRDP; MBD; Mbd1; MBD-1; MDP; membrane bound dipeptidase type II; membrane-bound dipeptidase 1; Microsomal dipeptidase; RDP; Renal dipeptidase; testicular tissue protein Li 57
Gene Symbols DPEP1
Host Species Mouse
Immunogen Recombinant DPEP1 (Asp17-Leu411) expressed in E.coli.
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1800
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.