Learn More
CPC Scientific H-Tyr-DPen-Gly-(4Cl)Phe-DPen-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific ENKP005B
Encompass
Highly selective ligand with high affinity for the delta-opioid receptor.SEQUENCE: H-Tyr-DPen-Gly-(4Cl)Phe-DPen-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: Y-DPen-G-(4Cl)Phe-DPenMOLECULAR FORMULA: C30H38N5O7S2Cl1MOLECULAR WEIGHT: 680.3STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: PainREFERENCES:L.K. Vaughn et al., Life Sciences, 45, 1001 (1989)SKU(s): ENKP-005A, ENKP-005B
Catalog No. 50-210-9257
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.