Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DPM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121135
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121135 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121135 Supplier Invitrogen™ Supplier No. PA5121135
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: NCI-H460, HeLa, SKOV3, Mouse pancreas, Mouse liver, Mouse kidney, Rat liver. The target is usually found in the following locations: Endoplasmic reticulum. Immunogen sequence: MASLEVSRSP RRSRRELEVR SPRQNKYSVL LPTYNERENL PLIVWLLVKS FSESGINYEI IIIDDGSPDG TRDVAEQLEK IYGSDRILLR PREKKLGLGT AYIHGMKHAT GNYIIIMDAD LSHHPKFIPE FIRKQKEGNF DIVSGTRYKG NGGVYGWDLK RKIISRGANF LTQILLRPGA SDLTGSFRLY RKEVLEKLIE KCVSKGYVFQ MEMIVRARQL NYTIGEVPIS FVDRVYGESK LGGNEIVSFL KGLLTLFATT.

Dolichol-phosphate mannose (Dol-P-Man) serves as a donor of mannosyl residues on the lumenal side of the endoplasmic reticulum (ER). Lack of Dol-P-Man results in defective surface expression of GPI-anchored proteins. Dol-P-Man is synthesized from GDP-mannose and dolichol-phosphate on the cytosolic side of the ER by the enzyme dolichyl-phosphate mannosyltransferase. Human DPM1 lacks a carboxy-terminal transmembrane domain and signal sequence and is regulated by DPM2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPM1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.44 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DPM1
Gene Accession No. O60762, O70152
Gene Alias AI118379; AI194292; CDGIE; dolichol monophosphate mannose synthase; dolichol-phosphate (beta-D) mannosyltransferase 1; Dolichol-phosphate mannose synthase; Dolichol-phosphate mannose synthase subunit 1; Dolichol-phosphate mannosyltransferase; Dolichol-phosphate mannosyltransferase subunit 1; dolichyl-phosphate beta-D-mannosyltransferase; Dolichyl-phosphate beta-D-mannosyltransferase subunit 1; dolichyl-phosphate mannosyltransferase 1; dolichyl-phosphate mannosyltransferase polypeptide 1 catalytic subunit; dolichyl-phosphate mannosyltransferase polypeptide 1, catalytic subunit; DPM synthase; DPM synthase complex, catalytic subunit; DPM synthase subunit 1; Dpm1; Mannose-P-dolichol synthase; Mannose-P-dolichol synthase subunit 1; MPD synthase; MPD synthase subunit 1; MPDS; OTTMUSP00000017164; P9705.3; RP23-391M18.1; SED3; YPR183W
Gene Symbols DPM1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human DPM1 (NP_003850.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13480, 296394, 8813
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.