Learn More
Abcam DPN, ERβ Agonist, Selective, Solid, 10 mg
Description
- Potency: Potent, selective ERβ agonist with EC50 values of 0.85 nM for ERβ and 66 nM for ERα.
- Purity: >99%.
- Molecular Weight: 239.27 Da.
- Form: Solid.
- Composition: 100% 2,3-Bis(4-hydroxphenyl)propionitrile.
- Applications: Demonstrated cardioprotective and neuroprotective effects in vivo.
- Storage: Store at -20°C.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Estrogen Receptor β |
| Shipping Condition | Ambient |
| Buffer | 100% 2,3-Bis(4-hydroxphenyl)propionitrile |
| Content And Storage | Store at -20°C. |
| Format | Vial |
| Form | Solid |
| Formulation | 100% 2,3-Bis(4-hydroxphenyl)propionitrile |
| Quantity | 10 mg |
| For Use With (Application) | Cardioprotection, Neuroprotection |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.