Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DPPA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112405 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112405 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112405 Supplier Invitrogen™ Supplier No. PA5112405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DPPA5 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Involved in the maintenance of embryonic stem (ES) cell pluripotency. Dispensable for self-renewal of pluripotent ES cells and establishment of germ cells. Associates with specific target mRNAs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPPA5
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene DPPA5
Gene Accession No. A6NC42, Q9CQS7
Gene Alias AA536857; developmental pluripotency associated 5; developmental pluripotency associated 5A; developmental pluripotency-associated 5 protein; Developmental pluripotency-associated protein 5A; Dppa5; Dppa5a; Ecat2; embryonal stem cell specific gene 1; embryonal stem cell-specific gene 1 protein; ES cell-associated transcript 2 protein; ESG1; ESG-1; ESG1ESG-1; hDPPA5; mDppa5; Ph34; Protein pH 34
Gene Symbols DPPA5, Dppa5a
Host Species Rabbit
Immunogen Recombinant Human Developmental pluripotency-associated 5 protein (1-116AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340168, 434423
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.