Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPY19L2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988438
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8438 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8438 Supplier Bioss Supplier No. BS8291R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Gene ontology (GO) annotation includes protein C-linked glycosylation via 2′-alpha-mannosyl-L-tryptophan; spermatid development.

Specifications

Antigen DPY19L2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DPY19L2
Gene Accession No. Q6NUT2
Gene Alias 4932443J21Rik; BOS_4293; dpy-19 like 2; DPY19L2; dpy-19-like 2; dpy-19-like 2 (C. elegans); Dpy-19-like protein 2; Dyp19l2; Gm18376; LOC524676; Probable C-mannosyltransferase DPY19L2; probable C-mannosyltransferase DPY19L2-like; Protein dpy-19 homolog 2; RGD1564311; SPATA34; spermatogenesis associated 34; SPGF9; UNQ3127/PRO10284
Gene Symbols DPY19L2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DPY19L2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283417
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.