Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DPYD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA522302
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522302 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522302 Supplier Invitrogen™ Supplier No. PA522302
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Raji, K562, NCI-H929. Predicted reactivity: Mouse (91%), Rat (92%), Xenopus laevis (91%), Pig (94%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Involved in pyrimidine base degradation. Catalyzes the reduction of uracil and thymine. Also involved the degradation of the chemotherapeutic drug 5-fluorouracil.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPYD
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.59 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene DPYD
Gene Accession No. Q12882
Gene Alias AI315208; DHP; DHPDHase; dihydropyrimidine dehydrogenase; dihydropyrimidine dehydrogenase [NADP(+)]; Dihydrothymine dehydrogenase; dihydrouracil dehydrogenase; DPD; Dpyd; E330028L06Rik
Gene Symbols DPYD
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human DPYD. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1806
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.