Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DPYS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112406
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112406 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112406 Supplier Invitrogen™ Supplier No. PA5112406
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DPYS Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Dihydropyrimidinase catalyzes the conversion of 5,6-dihydrouracil to 3-ureidopropionate in pyrimidine metabolism. Dihydropyrimidinase is expressed at a high level in liver and kidney as a major 2.5-kb transcript and a minor 3.8-kb transcript. Defects in the DPYS gene are linked to dihydropyrimidinuria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPYS
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Dpys
Gene Accession No. Q14117, Q9EQF5
Gene Alias 1200017I10Rik; 1300004I01Rik; DHP; DHPase; dihydropyrimidinase; Dihydropyrimidine amidohydrolase; Dpys; Hydantoinase
Gene Symbols Dpys
Host Species Rabbit
Immunogen Recombinant Human Dihydropyrimidinase protein (250-519AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1807, 64705
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.