Learn More
Dräger™ 7000 Tube Opener
Catalog No. 19810247 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Detector Tube Systems
Catalog No. For Use With (Equipment)
19-810-247 Detector Tube Systems
1 options
Catalog No. 19-810-247 Supplier Dräger™ Supplier No. 6401200
  • $136.00
    $117.65 / Each of 1
    Save $18.35
    13% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart
An accessory for detector tube systems.
  • Accessory for detector tube systems
  • For scoring and breaking tips off individual Draeger detector tubes
  • Glass tips safely stored for proper disposal
  • Aids in breaking tube ampoules
Specifications
For Use With (Equipment) Detector Tube Systems
Type 7000 Tube Opener