Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dräger™ XXS Replacement Ammonia DragerSensor™

Catalog No. 17985674 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Pac 7000, X-am 5000 and X-am 5600 Multi-Gas Monitors
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
17-985-674 Pac 7000, X-am 5000 and X-am 5600 Multi-Gas Monitors
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-985-674 Supplier Dräger™ Supplier No. 6810888
  • $993.00
    $712.65 / Each of 1

    Save $280.35

    28% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Pac 7000, X-am 5000 and X-am 5600 Multi-Gas Monitors.

  • Measuring Range: 0 to 300ppm
  • Resolution: 1ppm

Warranty and Services

One year

Specifications

Detection Limits 0 to 300ppm
Components Measured Ammonia
For Use With (Equipment) Pac 7000, X-am 5000 and X-am 5600 Multi-Gas Monitors
Type Ammonia Replacement Sensor
For Use With (Application) Accurate detection of Ammonia
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.