Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dräger™ Carbon Monoxide (hydrogen-compensated) (CO-H2) XXS Sensor

Catalog No. 17980844 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Components Measured:
Carbon Monoxide (Hydrogen-compensated)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
17-980-844 Carbon Monoxide (Hydrogen-compensated)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-980-844 Supplier Dräger™ Supplier No. 6811950
  • $1,002.00
    $729.65 / Each of 1

    Save $272.35

    27% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

XXS Replacement Sensor, for use with X-am 5000 multi-gas detector.

  • Measuring range: 0 to 2000 ppm
  • Resolution: 2 ppm
  • Carbon Monoxide (hydrogen-compensated) (CO-H2)

Specifications

Components Measured Carbon Monoxide (Hydrogen-compensated)
For Use With (Equipment) X-am 5000 Multigas Detector
Type Replacement Sensor
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.