Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dräger™ Carbon Monoxide/Hydrogen Sulfide (CO/H2S) XXS Replacement Sensor

Catalog No. 17980813 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Components Measured:
Carbon Monoxide, Hydrogen Sulfide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
17-980-813 Carbon Monoxide, Hydrogen Sulfide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-980-813 Supplier Dräger™ Supplier No. 6811410

missing translation for 'validMainframeMsg'

XXS Replacement Sensor, for use with X-am 5000 multi-gas detectors.

  • Measuring range: 0 to 2000 ppm (CO); 0-200 ppm (H2S)
  • Resolution: 2 ppm (CO); 1 ppm (H2S)
  • Carbon Monoxide (CO); Hydrogen Sulfide (H2S)

Specifications

Components Measured Carbon Monoxide, Hydrogen Sulfide
For Use With (Equipment) X-am 5000 Multigas Detector
Type Replacement Sensor
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.