Description
The semi-quantitative test takes four minutes to provide on the spot results
- No power supply required and the tubes are pre-calibrated
- On-the-spot results in four minutes
- No warm-up time
- No calibration
- Minimal training
- Simple operation
- Five-tube reagent set and laminated instruction card available separately; see cat. nos. 19080708 and 19080704
Semi-quantitative measurement of hazardous substances
Specifications
Specifications
| Type | Clandestine Lab Simultest Kit |
| Includes | Accuro™ pump, test set adapter and opener, quick reference laminated instruction sheet and 5 clandestine lab simultest set, Pelican™ case |