Description
Includes
Standard kit consists of an impact-resistant Pelican™ case with a custom-cut foam insert and labels, the CMS analyzer, the remote sampling system, the telescopic probe, and 10 chips. Kit is also available without chips, allowing the user to customize the kit for specific situations from Dräger's extensive selection of CMS chips, available on www.fishersafety.com.
For gas and vapor spot measurement.
- Analyzer uses substance-specific chips to obtain accurate, reliable results
- Chip contains an electronic bar code that defines the parameters to be tested, and a substance-specific reagent that reacts quickly with the contaminant
See Dräger CMS Chip Measure System (18-999-1915A) for details on system operation.
Specifications
Specifications
| Type | CMS Emergency Response Kit |