Learn More
Dräger™ CMS Chips: Acetone
Catalog No. 19040949 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
40 to 600
Catalog No. Measurement Range
19-040-949 40 to 600
1 options
Catalog No. 19-040-949 Supplier Dräger™ Supplier No. 6406470
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Acetone CMS Chip
Humidity Range 0 to 30mg/L
Measurement Range 40 to 600
Response Time 60 to 600 sec.
Temperature Range (English) 41°F to 104°F
Temperature Range (Metric) 5° to 40°C