Learn More
Dräger™ CMS Chips: Ethanol
Catalog No. 19640370 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
100 to 2500
Catalog No. Measurement Range
19-640-370 100 to 2500
1 options
Catalog No. 19-640-370 Supplier Dräger™ Supplier No. 6406370
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Ethanol CMS Chip
Humidity Range 5 to 25mg/L
Measurement Range 100 to 2500
Response Time 60 to 340 sec.
Temperature Range (English) 50°F to 86°F
Temperature Range (Metric) 10°C to 30°C