Learn More
Dräger™ CMS Chips: Isopropanol
Catalog No. 19640390 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
40 to 1000
Catalog No. Measurement Range
19-640-390 40 to 1000
1 options
Catalog No. 19-640-390 Supplier Dräger™ Supplier No. 6406390
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Isopropanol CMS Chip
Humidity Range 5 to 25mg/L
Measurement Range 40 to 1000
Response Time 100 to 550 sec.
Temperature Range (English) 50°F to 86°F
Temperature Range (Metric) 10°C to 30°C