Learn More
Dräger™ CMS Chips: Carbon Monoxide
Catalog No. 189991921 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
5 to 150
Catalog No. Measurement Range
18-999-1921 5 to 150
1 options
This item is not returnable. View return policy
Catalog No. 18-999-1921 Supplier Dräger™ Supplier No. 6406080
This item is not returnable. View return policy
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Carbon Monoxide CMS Chip
Humidity Range 1 to 50mg/L
Measurement Range 5 to 150
Response Time 80 to 300 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C