Learn More
Dräger™ CMS Chips: Benzene
Catalog No. 19034569 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
0.2 to 10
0.5 to 10
10 to 250
Response Time:
35 to 225 sec.
35 to 300 sec.
40 to 275 sec.
Humidity Range:
1 to 30mg/L
1 to 40mg/L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Range Response Time Humidity Range
19-034-569 10 to 250 40 to 275 sec. 1 to 30mg/L
19-815-818 0.2 to 10 35 to 300 sec. 1 to 30mg/L
19-034-568 0.5 to 10 35 to 225 sec. 1 to 40mg/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-034-569 Supplier Dräger™ Supplier No. 6406280
Suitable for 10 measurements of one gas
 Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Benzene CMS Chip
Humidity Range 1 to 30mg/L
Measurement Range 10 to 250
Response Time 40 to 275 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C