Learn More
Dräger™ CMS Chips: Carbon Dioxide
Catalog No. 189991920 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
1000 to 25000
Response Time:
25 to 140 sec.
Catalog No. Measurement Range Response Time
18-999-1920 1000 to 25000 25 to 140 sec.
1 options
Catalog No. 18-999-1920 Supplier Dräger™ Supplier No. 6406070
Suitable for 10 measurements of one gas
 Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Carbon Dioxide CMS Chip
Humidity Range 1 to 30mg/L
Measurement Range 1000 to 25000
Response Time 25 to 140 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C