Learn More
Dräger™ CMS Chips: Perchloroethylene
Catalog No. 189991930 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
5 to 500
Catalog No. Measurement Range
18-999-1930 5 to 500
1 options
Catalog No. 18-999-1930 Supplier Dräger™ Supplier No. 6406040
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Perchloroethylene CMS Chip
Humidity Range 5 to 30mg/L
Measurement Range 5 to 500
Response Time 40 to 330 sec.
Temperature Range (English) 59°F to 104°F
Temperature Range (Metric) 15° to 40°C