Learn More
Dräger™ CMS Chips: Ozone
Catalog No. 19040950 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
25 to 1000
Catalog No. Measurement Range
19-040-950 25 to 1000
1 options
This item is not returnable. View return policy
Catalog No. 19-040-950 Supplier Dräger™ Supplier No. 6406430
This item is not returnable. View return policy
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Ozone CMS Chip
Humidity Range 1 to 25mg/L
Measurement Range 25 to 1000
Response Time 100 to 600 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C