Learn More
Dräger™ CMS Chips: Training Chip
Catalog No. 19640290 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Humidity Range:
1 to 40 mg/L
Catalog No. Humidity Range
19-640-290 1 to 40 mg/L
1 options
This item is not returnable. View return policy
Catalog No. 19-640-290 Supplier Dräger™ Supplier No. 6406290
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Training Chip CMS Chip
Humidity Range 1 to 40 mg/L
Response Time 30 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C