Learn More
Dräger™ CMS Chips: Propane
Catalog No. 19640310 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
100 to 2000
Catalog No. Measurement Range
19-640-310 100 to 2000
1 options
Catalog No. 19-640-310 Supplier Dräger™ Supplier No. 6406310
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Propane CMS Chip
Humidity Range 1 to 40mg/L
Measurement Range 100 to 2000
Response Time 60 to 360 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C