Learn More
Dräger™ CMS Chips: Trichloroethylene
Catalog No. 19640320 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
5 to 100
Catalog No. Measurement Range
19-640-320 5 to 100
1 options
Catalog No. 19-640-320 Supplier Dräger™ Supplier No. 6406320
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Trichloroethylen CMS Chip
Humidity Range 1 to 30mg/L
Measurement Range 5 to 100
Response Time 40 to 330 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C