Learn More
Dräger™ CMS Chips: Phosgene
Catalog No. 19640340 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
0.05 to 2
Catalog No. Measurement Range
19-640-340 0.05 to 2
1 options
Catalog No. 19-640-340 Supplier Dräger™ Supplier No. 6406340
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Phosgene CMS Chip
Humidity Range 1 to 30mg/L
Measurement Range 0.05 to 2
Response Time 90 to 420 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C