Learn More
Dräger™ CMS Chips: Methanol
Catalog No. 19640380 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
20 to 500
Catalog No. Measurement Range
19-640-380 20 to 500
1 options
Catalog No. 19-640-380 Supplier Dräger™ Supplier No. 6406380
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Methanol CMS Chip
Humidity Range 5 to 25mg/L
Measurement Range 20 to 500
Response Time 200 to 600 sec.
Temperature Range (English) 59°F to 104°F
Temperature Range (Metric) 15°C to 30°C