Learn More
Dräger™ CMS Chips: Xylene
Catalog No. 19034579 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
10 to 300
Catalog No. Measurement Range
19-034-579 10 to 300
1 options
Catalog No. 19-034-579 Supplier Dräger™ Supplier No. 6406260
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type o-Xylene CMS Chip
Humidity Range 1 to 30mg/L
Measurement Range 10 to 300
Response Time 75 to 500 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C