Learn More
Dräger™ CMS Chips: MTBE
Catalog No. 17985357 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
10 to 200
Catalog No. Measurement Range
17-985-357 10 to 200
1 options
Catalog No. 17-985-357 Supplier Dräger™ Supplier No. 6406530
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type MTBE CMS Chip
Humidity Range 1 to 30mg/L
Measurement Range 10 to 200
Response Time 90 to 450 sec.
Temperature Range (English) 50°F to 86°F
Temperature Range (Metric) 10°C to 30°C