Learn More
Dräger™ CMS Chips: Nitrous Gases
Catalog No. 189991929 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
0.5 to 15
10 to 200
Response Time:
20 to 100 sec.
40 to 350 sec.
Humidity Range:
1 to 30mg/L
1 to 40mg/L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Range Response Time Humidity Range
18-999-1929 0.5 to 15 40 to 350 sec. 1 to 40mg/L
19-640-240 10 to 200 20 to 100 sec. 1 to 30mg/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-999-1929 Supplier Dräger™ Supplier No. 6406060
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Nitrous Gases CMS Chip
Humidity Range 1 to 40mg/L
Measurement Range 0.5 to 15
Response Time 40 to 350 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C