Learn More
Dräger™ CMS Chips: Sulfur Dioxide
Catalog No. 19034575 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
0.4 to 10
5 to 150
Response Time:
40 to 300 sec.
40 to 360 sec.
Humidity Range:
1 to 40mg/L
5 to 20mg/L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Range Response Time Humidity Range
19-034-575 5 to 150 40 to 360 sec. 1 to 40mg/L
18-999-1931 0.4 to 10 40 to 300 sec. 5 to 20mg/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-034-575 Supplier Dräger™ Supplier No. 6406180
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Sulfur Dioxide CMS Chip
Humidity Range 1 to 40mg/L
Measurement Range 5 to 150
Response Time 40 to 360 sec.
Temperature Range (English) 41°F to 104°F
Temperature Range (Metric) 5° to 40°C